Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGGGTCCGCATCACCCCTGGC[C/T]GGTCCGACAGACCGCCCGCCTCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600307 MIM: 604657 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLTPD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLTPD2 - glycolipid transfer protein domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014985.2 | 890 | Missense Mutation | CCG,CTG | P,L 229 | NP_001014985.2 | |
XM_005256626.3 | 890 | Missense Mutation | CCG,CTG | P,L 265 | XP_005256683.1 |
PSMB6 - proteasome subunit beta 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM4SF5 - transmembrane 4 L six family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VMO1 - vitelline membrane outer layer 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |