Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTCCCTGGGGCTCGTCCAGGC[C/T]CGGGCGCTGGCTGGGAGGGGAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHH3 - pleckstrin homology, MyTH4 and FERM domain containing H3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024927.4 | 2632 | Missense Mutation | AGC,GGC | S,G 777 | NP_079203.3 | |
XM_017025113.1 | 2632 | Missense Mutation | AGC,GGC | S,G 866 | XP_016880602.1 | |
XM_017025114.1 | 2632 | Missense Mutation | AGC,GGC | S,G 863 | XP_016880603.1 | |
XM_017025115.1 | 2632 | Missense Mutation | AGC,GGC | S,G 822 | XP_016880604.1 | |
XM_017025116.1 | 2632 | Missense Mutation | AGC,GGC | S,G 821 | XP_016880605.1 | |
XM_017025117.1 | 2632 | Missense Mutation | AGC,GGC | S,G 806 | XP_016880606.1 | |
XM_017025118.1 | 2632 | Missense Mutation | AGC,GGC | S,G 774 | XP_016880607.1 |
TUBG2 - tubulin gamma 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |