Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTATGTTGTCGAGCAAAGCAAAC[A/T]TCTGCCCCTGGTGTGGCAGAATACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607740 | ||||||||||||||||||||
Literature Links: |
SCARNA20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCARNA20 - small Cajal body-specific RNA 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP32 - ubiquitin specific peptidase 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032582.3 | 964 | Missense Mutation | GAA,GAT | E,D 488 | NP_115971.2 | |
XM_011525371.1 | 964 | Missense Mutation | GAA,GAT | E,D 504 | XP_011523673.1 | |
XM_011525372.1 | 964 | Missense Mutation | GAA,GAT | E,D 504 | XP_011523674.1 | |
XM_011525373.1 | 964 | Missense Mutation | GAA,GAT | E,D 488 | XP_011523675.1 | |
XM_011525374.1 | 964 | Missense Mutation | GAA,GAT | E,D 485 | XP_011523676.1 | |
XM_011525375.1 | 964 | Missense Mutation | GAA,GAT | E,D 478 | XP_011523677.1 | |
XM_011525376.1 | 964 | Missense Mutation | GAA,GAT | E,D 485 | XP_011523678.1 | |
XM_011525378.1 | 964 | Missense Mutation | GAA,GAT | E,D 442 | XP_011523680.1 | |
XM_011525379.2 | 964 | Missense Mutation | GAA,GAT | E,D 197 | XP_011523681.1 | |
XM_017025233.1 | 964 | Missense Mutation | GAA,GAT | E,D 442 | XP_016880722.1 | |
XM_017025234.1 | 964 | Missense Mutation | GAA,GAT | E,D 158 | XP_016880723.1 |