Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGTGCAGCCTGTGGAGCCTTC[A/G]CTGGGGGAAGCTCTGGGGTGGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614144 MIM: 607263 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B9D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B9D1 - B9 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243473.2 | 871 | UTR 3 | NP_001230402.1 | |||
NM_001243475.2 | 871 | Intron | NP_001230404.1 | |||
NM_001321214.1 | 871 | UTR 3 | NP_001308143.1 | |||
NM_001321215.1 | 871 | UTR 3 | NP_001308144.1 | |||
NM_001321216.1 | 871 | Intron | NP_001308145.1 | |||
NM_001321217.1 | 871 | Intron | NP_001308146.1 | |||
NM_001321218.1 | 871 | Intron | NP_001308147.1 | |||
NM_001321219.1 | 871 | Intron | NP_001308148.1 | |||
XM_005256607.2 | 871 | UTR 3 | XP_005256664.1 | |||
XM_005256610.1 | 871 | Intron | XP_005256667.1 | |||
XM_017024452.1 | 871 | Intron | XP_016879941.1 |
EPN2 - epsin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1180 - microRNA 1180 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |