Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAGGAGGAGGTCTCTGTTTGAT[A/G]AGCGTCGTTTGCATCTGAAAGTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604858 | ||||||||||||||||||||
Literature Links: |
SRP68 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRP68 - signal recognition particle 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260502.1 | 1650 | Missense Mutation | CAT,TAT | H,Y 501 | NP_001247431.1 | |
NM_001260503.1 | 1650 | Missense Mutation | CAT,TAT | H,Y 200 | NP_001247432.1 | |
NM_014230.3 | 1650 | Missense Mutation | CAT,TAT | H,Y 539 | NP_055045.2 |