Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTCAGGACAGAATGAGGGATAG[C/T]TCGGCCCCCAGCTCGGCCTCCTCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601934 MIM: 602578 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUS1L - dihydrouridine synthase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFNG - RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002917.1 | 131 | Intron | NP_002908.1 | |||
XM_011523587.2 | 131 | Intron | XP_011521889.1 |