Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTACCCCGGCCCCCAGTCGGC[C/T]GCCTCCGGCTTTCTTCCCCTCGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605339 MIM: 604041 MIM: 601297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FXR2 - FMR1 autosomal homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004860.3 | 1813 | Missense Mutation | CAG,CGG | Q,R 483 | NP_004851.2 |
LOC100996842 - uncharacterized LOC100996842 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPDU1 - mannose-P-dolichol utilization defect 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX15 - SRY-box 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |