Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAGAACTCGCACTGGTAGGGCC[C/G]CACGCCCGTGTGGATGCGCATGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616591 | ||||||||||||||||||||
Literature Links: |
ZBTB7C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB7C - zinc finger and BTB domain containing 7C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039360.2 | 2049 | Missense Mutation | CGG,GGG | R,G 446 | NP_001034449.1 | |
NM_001318841.1 | 2049 | Missense Mutation | CGG,GGG | R,G 446 | NP_001305770.1 | |
XM_005258229.4 | 2049 | Missense Mutation | CGG,GGG | R,G 446 | XP_005258286.1 | |
XM_011525861.2 | 2049 | Missense Mutation | CGG,GGG | R,G 495 | XP_011524163.1 | |
XM_011525863.2 | 2049 | Missense Mutation | CGG,GGG | R,G 455 | XP_011524165.1 | |
XM_011525864.2 | 2049 | Missense Mutation | CGG,GGG | R,G 455 | XP_011524166.1 | |
XM_011525865.2 | 2049 | Missense Mutation | CGG,GGG | R,G 455 | XP_011524167.1 | |
XM_011525866.2 | 2049 | Missense Mutation | CGG,GGG | R,G 455 | XP_011524168.1 | |
XM_011525867.2 | 2049 | Missense Mutation | CGG,GGG | R,G 455 | XP_011524169.1 | |
XM_011525869.2 | 2049 | Missense Mutation | CGG,GGG | R,G 454 | XP_011524171.1 | |
XM_011525870.2 | 2049 | Missense Mutation | CGG,GGG | R,G 446 | XP_011524172.1 | |
XM_011525871.2 | 2049 | Missense Mutation | CGG,GGG | R,G 446 | XP_011524173.1 | |
XM_017025605.1 | 2049 | Missense Mutation | CGG,GGG | R,G 495 | XP_016881094.1 | |
XM_017025606.1 | 2049 | Missense Mutation | CGG,GGG | R,G 455 | XP_016881095.1 | |
XM_017025607.1 | 2049 | Missense Mutation | CGG,GGG | R,G 455 | XP_016881096.1 | |
XM_017025608.1 | 2049 | Missense Mutation | CGG,GGG | R,G 454 | XP_016881097.1 | |
XM_017025609.1 | 2049 | Missense Mutation | CGG,GGG | R,G 446 | XP_016881098.1 |