Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCATGAGGGAAACTCTCACAAG[C/T]TAAGTAAGGAGGTGGATTAATTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614697 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDHD2 - haloacid dehalogenase like hydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318765.1 | 936 | Missense Mutation | ACT,GCT | T,A 153 | NP_001305694.1 | |
NM_032124.4 | 936 | Missense Mutation | ACT,GCT | T,A 243 | NP_115500.1 | |
XM_005258371.4 | 936 | Intron | XP_005258428.1 | |||
XM_006722557.2 | 936 | Missense Mutation | ACT,GCT | T,A 153 | XP_006722620.1 | |
XM_011526227.2 | 936 | Missense Mutation | ACT,GCT | T,A 243 | XP_011524529.1 | |
XM_011526228.2 | 936 | Missense Mutation | ACT,GCT | T,A 243 | XP_011524530.1 | |
XM_017026039.1 | 936 | Missense Mutation | ACT,GCT | T,A 243 | XP_016881528.1 | |
XM_017026040.1 | 936 | Intron | XP_016881529.1 |
KATNAL2 - katanin catalytic subunit A1 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |