Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAGTAGCGCGACACGTTGACCA[C/G]GTACTGCGGATGGAAGTAGTAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607162 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST8SIA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST8SIA5 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307986.1 | 1613 | Silent Mutation | CTG,GTG | L,V 318 | NP_001294915.1 | |
NM_001307987.1 | 1613 | Silent Mutation | CTG,GTG | L,V 251 | NP_001294916.1 | |
NM_013305.5 | 1613 | Silent Mutation | CTG,GTG | L,V 282 | NP_037437.2 | |
XM_017025715.1 | 1613 | Missense Mutation | CTG,GTG | L,V 208 | XP_016881204.1 | |
XM_017025716.1 | 1613 | Missense Mutation | CTG,GTG | L,V 134 | XP_016881205.1 |