Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAAGAGGATCCATCTGTTTCCA[A/G]TTATTGGCCTTTAACTGATGAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605445 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLGAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLGAP1 - DLG associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003809.2 | 1921 | Silent Mutation | AAC,AAT | N,N 596 | NP_001003809.1 | |
NM_001242761.1 | 1921 | Silent Mutation | AAC,AAT | N,N 898 | NP_001229690.1 | |
NM_001242762.1 | 1921 | Silent Mutation | AAC,AAT | N,N 582 | NP_001229691.1 | |
NM_001242763.1 | 1921 | Silent Mutation | AAC,AAT | N,N 620 | NP_001229692.1 | |
NM_001242764.1 | 1921 | Silent Mutation | AAC,AAT | N,N 604 | NP_001229693.1 | |
NM_001242765.1 | 1921 | Silent Mutation | AAC,AAT | N,N 596 | NP_001229694.1 | |
NM_001242766.1 | 1921 | Silent Mutation | AAC,AAT | N,N 606 | NP_001229695.1 | |
NM_001308390.1 | 1921 | Silent Mutation | AAC,AAT | N,N 614 | NP_001295319.1 | |
NM_004746.3 | 1921 | Silent Mutation | AAC,AAT | N,N 898 | NP_004737.2 | |
XM_005258171.2 | 1921 | Silent Mutation | AAC,AAT | N,N 908 | XP_005258228.1 | |
XM_005258172.1 | 1921 | Silent Mutation | AAC,AAT | N,N 908 | XP_005258229.1 | |
XM_005258173.3 | 1921 | Silent Mutation | AAC,AAT | N,N 685 | XP_005258230.2 | |
XM_005258174.4 | 1921 | Silent Mutation | AAC,AAT | N,N 616 | XP_005258231.1 | |
XM_006722367.3 | 1921 | Silent Mutation | AAC,AAT | N,N 652 | XP_006722430.1 | |
XM_011525770.2 | 1921 | Intron | XP_011524072.1 | |||
XM_011525771.2 | 1921 | Intron | XP_011524073.1 | |||
XM_017026080.1 | 1921 | Silent Mutation | AAC,AAT | N,N 642 | XP_016881569.1 | |
XM_017026081.1 | 1921 | Silent Mutation | AAC,AAT | N,N 610 | XP_016881570.1 | |
XM_017026082.1 | 1921 | Silent Mutation | AAC,AAT | N,N 675 | XP_016881571.1 | |
XM_017026083.1 | 1921 | Silent Mutation | AAC,AAT | N,N 606 | XP_016881572.1 | |
XM_017026084.1 | 1921 | Silent Mutation | AAC,AAT | N,N 606 | XP_016881573.1 | |
XM_017026085.1 | 1921 | Silent Mutation | AAC,AAT | N,N 604 | XP_016881574.1 |