Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGAACACGCTGACGGTGAAGTC[A/G]CTGGTCTCCGAGCCATACTTGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603508 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYOM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYOM1 - myomesin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003803.3 | 5032 | Silent Mutation | AGC,AGT | S,S 1657 | NP_003794.3 | |
NM_019856.1 | 5032 | Silent Mutation | AGC,AGT | S,S 1561 | NP_062830.1 | |
XM_017026062.1 | 5032 | Silent Mutation | AGC,AGT | S,S 1645 | XP_016881551.1 |