Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGCGAGCTGGCCGGAGCCCCCCG[A/C]CCCGGAGCCCGGGCCGCTGTTCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALR1 - galanin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001480.3 | 556 | Missense Mutation | GAC,GCC | D,A 20 | NP_001471.2 | |
XM_017025691.1 | 556 | Missense Mutation | GAC,GCC | D,A 20 | XP_016881180.1 |