Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGCTCAGCCCCAACTTCGACGC[G/T]CACCACACTGCCATGCTGACCCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604894 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ONECUT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ONECUT2 - one cut homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004852.2 | 1089 | Silent Mutation | GCG,GCT | A,A 264 | NP_004843.2 | |
XM_017026096.1 | 1089 | Silent Mutation | GCG,GCT | A,A 264 | XP_016881585.1 |