Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAAGTGTTTTAGTTCTGGAATA[A/G]CACATTAGTGGAGCCTGTTGAAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609674 | ||||||||||||||||||||
Literature Links: |
ESCO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESCO1 - establishment of sister chromatid cohesion N-acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052911.2 | 3040 | Silent Mutation | TGC,TGT | C,C 702 | NP_443143.2 | |
XM_011525798.1 | 3040 | Silent Mutation | TGC,TGT | C,C 702 | XP_011524100.1 | |
XM_011525799.2 | 3040 | Silent Mutation | TGC,TGT | C,C 672 | XP_011524101.1 |
GREB1L - growth regulation by estrogen in breast cancer 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |