Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATGGGGGAAGCAGAGAAGTTTCA[C/T]TACATCTATAGTTGTGACCTGGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3C3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3C3 - phosphatidylinositol 3-kinase catalytic subunit type 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308020.1 | 118 | Silent Mutation | CAC,CAT | H,H 8 | NP_001294949.1 | |
NM_002647.3 | 118 | Silent Mutation | CAC,CAT | H,H 8 | NP_002638.2 | |
XM_011526031.2 | 118 | UTR 5 | XP_011524333.1 |