Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACGTGAAAATCCGCAGCAGGAAG[C/T]TTGGGGTGAGTGGCTCGCTCGGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611402 | ||||||||||||||||||||
Literature Links: |
DOK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DOK6 - docking protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152721.5 | 258 | Missense Mutation | CTT,TTT | L,F 21 | NP_689934.2 | |
XM_011525875.2 | 258 | Missense Mutation | CTT,TTT | L,F 21 | XP_011524177.1 | |
XM_017025610.1 | 258 | Intron | XP_016881099.1 | |||
XM_017025611.1 | 258 | Intron | XP_016881100.1 |