Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATCCAGTTTCCTGCTAATTTGC[A/T]GCTTCCTCCAGGTATGTTGATAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601689 | ||||||||||||||||||||
Literature Links: |
TAF4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAF4B - TATA-box binding protein associated factor 4b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293725.1 | 383 | Missense Mutation | CAG,CTG | Q,L 111 | NP_001280654.1 | |
NM_005640.2 | 383 | Missense Mutation | CAG,CTG | Q,L 111 | NP_005631.1 | |
XM_005258339.3 | 383 | Missense Mutation | CAG,CTG | Q,L 111 | XP_005258396.1 | |
XM_011526153.2 | 383 | Missense Mutation | CAG,CTG | Q,L 111 | XP_011524455.1 | |
XM_017025932.1 | 383 | Missense Mutation | CAG,CTG | Q,L 111 | XP_016881421.1 | |
XM_017025933.1 | 383 | UTR 5 | XP_016881422.1 |