Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCTGAAGACACCAGAGAGAAGG[A/G]CTGCAAACCCAAATCATGACATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C18orf21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C18orf21 - chromosome 18 open reading frame 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201474.1 | 613 | Missense Mutation | ACT,GCT | T,A 56 | NP_001188403.1 | |
NM_001201475.1 | 613 | Missense Mutation | ACT,GCT | T,A 56 | NP_001188404.1 | |
NM_001201476.1 | 613 | Intron | NP_001188405.1 | |||
NM_031446.4 | 613 | Missense Mutation | ACT,GCT | T,A 144 | NP_113634.3 | |
XM_005258364.4 | 613 | Missense Mutation | ACT,GCT | T,A 150 | XP_005258421.1 |
RPRD1A - regulation of nuclear pre-mRNA domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |