Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTTAATCAAAGATGTAGATGG[C/T]GGAATGGACCAGGACATATTTGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600389 | ||||||||||||||||||||
Literature Links: |
MEP1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEP1B - meprin A subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308171.1 | 125 | Silent Mutation | GGC,GGT | G,G 31 | NP_001295100.1 | |
NM_005925.2 | 125 | Silent Mutation | GGC,GGT | G,G 31 | NP_005916.2 | |
XM_011526013.2 | 125 | Silent Mutation | GGC,GGT | G,G 31 | XP_011524315.1 | |
XM_011526014.2 | 125 | Silent Mutation | GGC,GGT | G,G 31 | XP_011524316.1 |