Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGCTCATCTCCCCGCGTCCGG[C/T]TTGGATATCTTCGGGGACCTGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC11C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC11C - SEC11 homolog C, signal peptidase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307941.1 | 137 | Silent Mutation | GGC,GGT | G,G 15 | NP_001294870.1 | |
NM_033280.3 | 137 | Silent Mutation | GGC,GGT | G,G 15 | NP_150596.1 | |
XM_011526260.2 | 137 | Intron | XP_011524562.1 | |||
XM_017026073.1 | 137 | Intron | XP_016881562.1 |