Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTATCACTCATGATGAGGACAA[A/G]AGGGCTGCGTGCTGGGAAACATGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605234 | ||||||||||||||||||||
Literature Links: |
VN1R1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VN1R1 - vomeronasal 1 receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020633.3 | 1220 | Missense Mutation | CTT,TTT | L,F 323 | NP_065684.1 |
ZNF749 - zinc finger protein 749 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF772 - zinc finger protein 772 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |