Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGGCGTCTTCTTCCTGGGGAGTA[C/T]GACTTGGACCGGGACCGCGACTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf44 - chromosome 19 open reading frame 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288834.1 | 2486 | Intron | NP_001275763.1 | |||
NM_032207.3 | 2486 | UTR 3 | NP_115583.1 | |||
XM_006722920.3 | 2486 | UTR 3 | XP_006722983.1 | |||
XM_011528354.2 | 2486 | Intron | XP_011526656.1 | |||
XM_017027350.1 | 2486 | Intron | XP_016882839.1 |
CHERP - calcium homeostasis endoplasmic reticulum protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006387.5 | 2486 | Silent Mutation | NP_006378.3 |