Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCGGAAGAGGCAGCGCGTGCGA[C/G]CGAGAACCCGGAGCAGGTGGCGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611823 | ||||||||||||||||||||
Literature Links: |
MRPL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL4 - mitochondrial ribosomal protein L4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015956.2 | 249 | Missense Mutation | ACC,AGC | T,S 32 | NP_057040.2 | |
NM_146387.1 | 249 | Missense Mutation | ACC,AGC | T,S 32 | NP_666499.1 | |
NM_146388.1 | 249 | Missense Mutation | ACC,AGC | T,S 32 | NP_666500.1 | |
XM_011528045.2 | 249 | Missense Mutation | ACC,AGC | T,S 32 | XP_011526347.1 |