Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGTTCAGAGAGTTTGGCTGGGC[A/G]TGCCAGGCGCAACAGCATGATGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602652 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK6 - kallikrein related peptidase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012964.2 | 781 | Missense Mutation | CGC,TGC | R,C 114 | NP_001012982.1 | |
NM_001012965.2 | 781 | Missense Mutation | CGC,TGC | R,C 7 | NP_001012983.1 | |
NM_001319948.1 | 781 | Missense Mutation | CGC,TGC | R,C 7 | NP_001306877.1 | |
NM_001319949.1 | 781 | Missense Mutation | CGC,TGC | R,C 7 | NP_001306878.1 | |
NM_002774.3 | 781 | Missense Mutation | CGC,TGC | R,C 114 | NP_002765.1 |