Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCCCCGCAGGCCACTTCCCAAA[A/C]AAGCCACTGCCATCGGCGGGCACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605414 MIM: 602373 | ||||||||||||||||||||
Literature Links: |
ABCA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCA7 - ATP binding cassette subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019112.3 | 392 | Missense Mutation | AAA,AAC | K,N 58 | NP_061985.2 | |
XM_006722616.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_006722679.1 | |
XM_006722617.2 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_006722680.1 | |
XM_006722618.2 | 392 | Intron | XP_006722681.1 | |||
XM_011527628.2 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_011525930.1 | |
XM_011527629.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_011525931.1 | |
XM_011527630.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_011525932.1 | |
XM_011527631.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_011525933.1 | |
XM_011527632.1 | 392 | Intron | XP_011525934.1 | |||
XM_011527633.2 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_011525935.1 | |
XM_011527634.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_011525936.1 | |
XM_011527635.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_011525937.1 | |
XM_011527636.2 | 392 | Intron | XP_011525938.1 | |||
XM_017026142.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_016881631.1 | |
XM_017026143.1 | 392 | Missense Mutation | AAA,AAC | K,N 58 | XP_016881632.1 |
CNN2 - calponin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |