Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTCTCCCGTACTGCTCAAAAGA[C/T]ACGTAGCGGATGCCCTTGCCAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609094 MIM: 603021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO17 - F-box protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024907.6 | 870 | Silent Mutation | GTA,GTG | V,V 246 | NP_079183.4 | |
NM_148169.2 | 870 | Silent Mutation | GTA,GTG | V,V 255 | NP_680474.1 |
MRPS12 - mitochondrial ribosomal protein S12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |