Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCGCGGGTTGGGGGACCCTCCT[C/T]GCCCAGGCTTCGGCCCCCACTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORCS8-MEF2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BORCS8-MEF2B - BORCS8-MEF2B readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005919.3 | 1516 | Silent Mutation | GCA,GCG | A,A 263 | NP_005910.1 |
MEF2B - myocyte enhancer factor 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145785.1 | 1516 | Missense Mutation | AAG,GAG | K,E 301 | NP_001139257.1 |
TMEM161A - transmembrane protein 161A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |