Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCAGGGAGCGCGGGCGGCTCCT[A/G]GGCGGCGCCAGACTGCGGTGAGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605377 MIM: 600963 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHMG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BHMG1 - basic helix-loop-helix and HMG-box containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DMPK - dystrophia myotonica protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081560.2 | 2442 | Missense Mutation | CCA,CTA | P,L 618 | NP_001075029.1 | |
NM_001081562.2 | 2442 | Nonsense Mutation | CAG,TAG | Q,* 617 | NP_001075031.1 | |
NM_001081563.2 | 2442 | Missense Mutation | CCA,CTA | P,L 633 | NP_001075032.1 | |
NM_001288764.1 | 2442 | Missense Mutation | CCA,CTA | P,L 649 | NP_001275693.1 | |
NM_001288765.1 | 2442 | Nonsense Mutation | CAG,TAG | Q,* 533 | NP_001275694.1 | |
NM_001288766.1 | 2442 | UTR 3 | NP_001275695.1 | |||
NM_004409.4 | 2442 | Missense Mutation | CCA,CTA | P,L 623 | NP_004400.4 |
SIX5 - SIX homeobox 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |