Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTGGAGTCTGTTCCAGAAACAG[A/G]ACCACATGAAGAGTGGTCTTGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194555 | ||||||||||||||||||||
Literature Links: |
ZNF224 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF224 - zinc finger protein 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF284 - zinc finger protein 284 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011526907.2 | 595 | Missense Mutation | GAA,GGA | E,G 93 | XP_011525209.1 | |
XM_011526908.2 | 595 | Missense Mutation | GAA,GGA | E,G 93 | XP_011525210.1 |