Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAGGGCGCAGGGCAGACAAAAG[A/G]TGAGGGCCGCCCAGTCGTGCCCGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611764 MIM: 604267 | ||||||||||||||||||||
Literature Links: |
CNFN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNFN - cornifelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032488.3 | 336 | Missense Mutation | ACC,ATC | T,I 94 | NP_115877.2 | |
XM_005259332.2 | 336 | Missense Mutation | ACC,ATC | T,I 107 | XP_005259389.1 | |
XM_011527396.1 | 336 | Missense Mutation | ACC,ATC | T,I 107 | XP_011525698.1 | |
XM_011527397.1 | 336 | Missense Mutation | ACC,ATC | T,I 107 | XP_011525699.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101930071 - uncharacterized LOC101930071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEGF8 - multiple EGF like domains 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |