Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCACAGACCGCCTGCTGCACCTC[C/T]TGGGGCTGGAGAAGACGGCGTTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611495 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4F22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP4F22 - cytochrome P450 family 4 subfamily F member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173483.3 | 213 | Silent Mutation | CTG,TTG | L,L 12 | NP_775754.2 | |
XM_011527692.2 | 213 | Silent Mutation | CTG,TTG | L,L 12 | XP_011525994.1 | |
XM_011527693.2 | 213 | Silent Mutation | CTG,TTG | L,L 12 | XP_011525995.1 |