Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTCTCCTCCCTAACATGGTAA[C/T]TGCCGCTCCGTTGGTGCTCCCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146631 MIM: 609950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICAM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ICAM3 - intercellular adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320605.1 | 2090 | Missense Mutation | AAT,AGT | N,S 422 | NP_001307534.1 | |
NM_001320606.1 | 2090 | Missense Mutation | AAT,AGT | N,S 441 | NP_001307535.1 | |
NM_001320608.1 | 2090 | Missense Mutation | AAT,AGT | N,S 257 | NP_001307537.1 | |
NM_002162.4 | 2090 | Missense Mutation | AAT,AGT | N,S 518 | NP_002153.2 |
RAVER1 - ribonucleoprotein, PTB binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133452.2 | 2090 | Intron | NP_597709.2 | |||
XM_011527671.1 | 2090 | Intron | XP_011525973.1 |