Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTTGGACTGATCCGCACAGGTC[C/T]GACCTCCAGTTTCAGCCCGAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615567 MIM: 606476 | ||||||||||||||||||||
Literature Links: |
COQ8B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COQ8B - coenzyme Q8B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142555.2 | 489 | Intron | NP_001136027.1 | |||
NM_024876.3 | 489 | Intron | NP_079152.3 |
ITPKC - inositol-trisphosphate 3-kinase C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025194.2 | 489 | Silent Mutation | TCC,TCT | S,S 152 | NP_079470.1 | |
XM_017027324.1 | 489 | Intron | XP_016882813.1 |