Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCACTTAGAAAATCCTCTTCC[G/T]CTTCAGGTAGGAGTCCGCGTAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614585 MIM: 609950 MIM: 611639 | ||||||||||||||||||||
Literature Links: |
FDX1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FDX1L - ferredoxin 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAVER1 - ribonucleoprotein, PTB binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133452.2 | 2334 | Silent Mutation | AGG,CGG | R,R 752 | NP_597709.2 | |
XM_011527671.1 | 2334 | Silent Mutation | AGG,CGG | R,R 727 | XP_011525973.1 |
ZGLP1 - zinc finger, GATA-like protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |