Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTTGTAGAAGACCACCTGCCC[A/G]TCCTGCGGGTGCCCGGGAGCCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138322 MIM: 615729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPX4 - glutathione peroxidase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBNO2 - strawberry notch homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100122.1 | 3340 | Silent Mutation | GAC,GAT | D,D 977 | NP_001093592.1 | |
NM_014963.2 | 3340 | Silent Mutation | GAC,GAT | D,D 1034 | NP_055778.2 | |
XM_011527804.2 | 3340 | Silent Mutation | GAC,GAT | D,D 1034 | XP_011526106.1 |