Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCACCAGGCACCTAGTGGTTT[A/G]TGTTGGAGTTCCTCAGCTTCCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDR87 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR87 - WD repeat domain 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291088.1 | 7817 | Silent Mutation | CAC,CAT | H,H 2521 | NP_001278017.1 | |
NM_031951.4 | 7817 | Silent Mutation | CAC,CAT | H,H 2482 | NP_114157.4 | |
XM_011527360.2 | 7817 | Silent Mutation | CAC,CAT | H,H 2521 | XP_011525662.1 |