Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATAAAATGTGTGTTGTAGTCC[A/G]CAGAGAGATAAATGGTGAGTGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP8 - NLR family pyrin domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317000.1 | 424 | Missense Mutation | CAC,CGC | H,R 118 | NP_001303929.1 | |
NM_176811.2 | 424 | Missense Mutation | CAC,CGC | H,R 118 | NP_789781.2 |