Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCACCTCCTCATCCTCAGCCTC[A/G]GGGGCCGGGGCCTTCTGTTTGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GP6 - glycoprotein VI platelet | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RDH13 - retinol dehydrogenase 13 (all-trans/9-cis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145971.1 | 1079 | Silent Mutation | CCC,CCT | P,P 298 | NP_001139443.1 | |
NM_138412.3 | 1079 | Silent Mutation | CCC,CCT | P,P 227 | NP_612421.1 | |
XM_005258473.1 | 1079 | Silent Mutation | CCC,CCT | P,P 298 | XP_005258530.1 | |
XM_011526407.1 | 1079 | Silent Mutation | CCC,CCT | P,P 298 | XP_011524709.1 | |
XM_011526408.2 | 1079 | Intron | XP_011524710.1 |