Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAACGGACGTCTCCAATGCACC[A/G]CACAGGAACGGGCCCCCAGAGCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601983 MIM: 180901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372397 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105372397 - uncharacterized LOC105372397 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP4K1 - mitogen-activated protein kinase kinase kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042600.2 | 2012 | Silent Mutation | TGC,TGT | C,C 624 | NP_001036065.1 | |
NM_007181.5 | 2012 | Silent Mutation | TGC,TGT | C,C 624 | NP_009112.1 | |
XM_011526403.1 | 2012 | Silent Mutation | TGC,TGT | C,C 664 | XP_011524705.1 | |
XM_011526404.1 | 2012 | Silent Mutation | TGC,TGT | C,C 664 | XP_011524706.1 | |
XM_017026231.1 | 2012 | Silent Mutation | TGC,TGT | C,C 651 | XP_016881720.1 |
RYR1 - ryanodine receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |