Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAAAGAAAGGAGATTTGGGATA[C/T]TGTTCTGCCCCACTTAGTGATGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
VN1R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VN1R2 - vomeronasal 1 receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173856.2 | 828 | Silent Mutation | TAC,TAT | Y,Y 248 | NP_776255.2 |
VN1R4 - vomeronasal 1 receptor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF677 - zinc finger protein 677 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |