Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGTCCATGCGGGACTGGGTGTG[A/G]TCCATCTCTTGGGCGAAGGCATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610672 MIM: 603765 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IER2 - immediate early response 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NACC1 - nucleus accumbens associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX10 - syntaxin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271609.1 | 810 | Missense Mutation | ACC,ATC | T,I 202 | NP_001258538.1 | |
NM_001271610.1 | 810 | Silent Mutation | GAC,GAT | D,D 203 | NP_001258539.1 | |
NM_001271611.1 | 810 | Silent Mutation | GAC,GAT | D,D 154 | NP_001258540.1 | |
NM_003765.2 | 810 | Silent Mutation | GAC,GAT | D,D 203 | NP_003756.1 | |
XM_011528397.2 | 810 | Intron | XP_011526699.1 |