Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGATGCCTAGTGAGATGTGAGC[A/G]CTGCTTAAAAGCTTTGCCACACTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF677 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF677 - zinc finger protein 677 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317998.1 | 1655 | Missense Mutation | CGC,TGC | R,C 415 | NP_001304927.1 | |
NM_182609.3 | 1655 | Missense Mutation | CGC,TGC | R,C 415 | NP_872415.1 | |
XM_005258862.4 | 1655 | Missense Mutation | CGC,TGC | R,C 379 | XP_005258919.1 | |
XM_011526912.2 | 1655 | Missense Mutation | CGC,TGC | R,C 415 | XP_011525214.1 |