Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTATCATGGATCACCCAAGCATG[C/T]GCATCTTGTCTTCCTGCTGCAAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614770 MIM: 601717 MIM: 610850 | ||||||||||||||||||||
Literature Links: |
PCP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCP2 - Purkinje cell protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PET100 - PET100 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STXBP2 - syntaxin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127396.2 | 164 | Missense Mutation | CGC,TGC | R,C 39 | NP_001120868.1 | |
NM_001272034.1 | 164 | Missense Mutation | CGC,TGC | R,C 39 | NP_001258963.1 | |
NM_006949.3 | 164 | Missense Mutation | CGC,TGC | R,C 39 | NP_008880.2 | |
XM_011528210.1 | 164 | Missense Mutation | CGC,TGC | R,C 39 | XP_011526512.1 | |
XM_011528212.2 | 164 | Missense Mutation | CGC,TGC | R,C 39 | XP_011526514.1 |
XAB2 - XPA binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |