Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGATGAGGTGGTCACAGGTACA[C/T]GGGCTGCTCCTGGCCTGTAAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604262 MIM: 612689 | ||||||||||||||||||||
Literature Links: |
APBA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APBA3 - amyloid beta precursor protein binding family A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004886.3 | 2020 | Missense Mutation | ATG,GTG | M,V 573 | NP_004877.1 | |
XM_006722950.3 | 2020 | Silent Mutation | CCA,CCG | P,P 607 | XP_006723013.1 | |
XM_006722951.3 | 2020 | Silent Mutation | CCA,CCG | P,P 365 | XP_006723014.1 |
TJP3 - tight junction protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267560.1 | 2020 | Intron | NP_001254489.1 | |||
NM_001267561.1 | 2020 | Intron | NP_001254490.1 |