Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCTTCCGGCGCAGTGTGGTCC[A/G]TGGTGGGGCCAGGCGCTATGCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605631 MIM: 600380 MIM: 174761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAPSA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAPSA - napsin A aspartic peptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR1H2 - nuclear receptor subfamily 1 group H member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256647.1 | 631 | Intron | NP_001243576.1 | |||
NM_007121.5 | 631 | Missense Mutation | CAT,CGT | H,R 117 | NP_009052.3 |
POLD1 - DAN polymerase delta 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |