Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTGTCACCATTTTGAATTGTCT[A/G]TTGTTTAAAAAGGCATGGTGTATTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608640 | ||||||||||||||||||||
Literature Links: |
ZNF461 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF461 - zinc finger protein 461 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297623.2 | 916 | Nonsense Mutation | CAG,TAG | Q,* 214 | NP_001284552.1 | |
NM_001322821.1 | 916 | Nonsense Mutation | CAG,TAG | Q,* 231 | NP_001309750.1 | |
NM_001322823.1 | 916 | Nonsense Mutation | CAG,TAG | Q,* 206 | NP_001309752.1 | |
NM_001322825.1 | 916 | Nonsense Mutation | CAG,TAG | Q,* 206 | NP_001309754.1 | |
NM_001322826.1 | 916 | Nonsense Mutation | CAG,TAG | Q,* 183 | NP_001309755.1 | |
NM_001322827.1 | 916 | Nonsense Mutation | CAG,TAG | Q,* 183 | NP_001309756.1 | |
NM_001322828.1 | 916 | Nonsense Mutation | CAG,TAG | Q,* 149 | NP_001309757.1 | |
NM_153257.4 | 916 | Nonsense Mutation | CAG,TAG | Q,* 237 | NP_694989.2 | |
XM_011527486.2 | 916 | Nonsense Mutation | CAG,TAG | Q,* 110 | XP_011525788.1 | |
XM_017027470.1 | 916 | Nonsense Mutation | CAG,TAG | Q,* 193 | XP_016882959.1 |