Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCATTATGACTTCTTCACACT[C/T]AGCGCTTTGACATGGATCTTCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF563 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF563 - zinc finger protein 563 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145276.2 | 557 | Missense Mutation | AAG,GAG | K,E 107 | NP_660319.1 | |
XM_005259750.4 | 557 | Intron | XP_005259807.1 | |||
XM_005259751.3 | 557 | Intron | XP_005259808.1 | |||
XM_006722650.3 | 557 | Intron | XP_006722713.1 | |||
XM_006722651.3 | 557 | Intron | XP_006722714.1 | |||
XM_011527698.1 | 557 | Intron | XP_011526000.1 | |||
XM_011527699.2 | 557 | Missense Mutation | AAG,GAG | K,E 70 | XP_011526001.1 | |
XM_011527700.2 | 557 | Intron | XP_011526002.1 | |||
XM_017026332.1 | 557 | Intron | XP_016881821.1 |