Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATGGACAGCATAAGCACAGCC[A/T]TCTTACTCCTGCTCCTGGCTCTCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP2F1 - cytochrome P450 family 2 subfamily F member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000774.4 | 234 | Missense Mutation | ATC,TTC | I,F 8 | NP_000765.2 | |
XM_011526551.2 | 234 | Missense Mutation | ATC,TTC | I,F 8 | XP_011524853.1 | |
XM_011526552.2 | 234 | Missense Mutation | ATC,TTC | I,F 8 | XP_011524854.1 | |
XM_011526553.2 | 234 | Missense Mutation | ATC,TTC | I,F 83 | XP_011524855.1 | |
XM_011526554.2 | 234 | Intron | XP_011524856.1 | |||
XM_011526555.2 | 234 | Missense Mutation | ATC,TTC | I,F 83 | XP_011524857.1 | |
XM_017026384.1 | 234 | Missense Mutation | ATC,TTC | I,F 83 | XP_016881873.1 | |
XM_017026385.1 | 234 | Missense Mutation | ATC,TTC | I,F 83 | XP_016881874.1 | |
XM_017026386.1 | 234 | Missense Mutation | ATC,TTC | I,F 83 | XP_016881875.1 | |
XM_017026387.1 | 234 | Missense Mutation | ATC,TTC | I,F 83 | XP_016881876.1 |