Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGCAGGCTCCATGGCGGCTGGC[G/T]TCTTCCAGGTATGGCCCCGGCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611670 MIM: 607391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISYNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISYNA1 - inositol-3-phosphate synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSBP4 - single stranded DNA binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009998.3 | 710 | Intron | NP_001009998.1 | |||
NM_032627.4 | 710 | Intron | NP_116016.1 | |||
XM_005259790.3 | 710 | Missense Mutation | GTC,TTC | V,F 143 | XP_005259847.1 | |
XM_006722665.3 | 710 | Missense Mutation | GTC,TTC | V,F 143 | XP_006722728.1 | |
XM_006722666.2 | 710 | Intron | XP_006722729.1 | |||
XM_006722668.2 | 710 | Intron | XP_006722731.1 | |||
XM_017026431.1 | 710 | Missense Mutation | GTC,TTC | V,F 143 | XP_016881920.1 | |
XM_017026432.1 | 710 | Missense Mutation | GTC,TTC | V,F 132 | XP_016881921.1 | |
XM_017026433.1 | 710 | Intron | XP_016881922.1 | |||
XM_017026434.1 | 710 | Missense Mutation | GTC,TTC | V,F 143 | XP_016881923.1 | |
XM_017026435.1 | 710 | Missense Mutation | GTC,TTC | V,F 132 | XP_016881924.1 | |
XM_017026436.1 | 710 | Intron | XP_016881925.1 | |||
XM_017026437.1 | 710 | Missense Mutation | GTC,TTC | V,F 143 | XP_016881926.1 |